LABOPORT™ UN860 Vacuum Diaphragm Pumps, KNF
Fournisseur: KNF Neuberger Inc.
Certificats
A propos de cet article
Quiet, reliable operation without the hazards associated with handling messy pump oils or contaminated wastewater generated by aspirators.
- Handle corrosive and aggressive vapors without damage
- Oil-free
- End vacuums to 1.5 torr
- Feature solid PTFE heads and diaphragms to protect against corrosives and aggressive vapors
- Multiport Kalrez™ valves withstand the ingestion of condensed vapors without the risk of damaging the pump or degrading performance
Recommended for rotary evaporators, vacuum concentrators, gel dryers, vacuum ovens, and general vacuum applications.
Spécifications
- Catalog No:
- 89209-764
- 89209-762
- Model:
- N860.1.2FTP
- N860.3FTP
- Vacuum:
- 75 torr
- 1.5 torr
- Protection class:
- IP 20 enclosure rating
- IP 20 enclosure rating
- W×D×H:
- 331×291×278 mm
- 331×291×278 mm