LABOPORT™ PowerDry™ Vacuum Diaphragm Pumps, Models UN820, UN840, UN842 and UN860, KNF
Fournisseur: KNF Neuberger Inc.
Certificats
A propos de cet article
Chemically resistant series of Laborport™ diaphragm vacuum pumps with the KNF PowerDry™ system that allows condensed liquids to be blown out of the pump heads at high speed during evacuation. These are best when moist gases must be pumped down such as when using a vacuum oven.
- Maintains vacuum levels by closing the pump suction valve during the purge cycle
- Integrated controller cycle times are factory set for optimum performance and are field adjustable
- Premium, corrosion-resistant PTFE and Kalrez™ resins used throughout
- Oil-free for contamination-free pumping
- Easily disassembled for occasional cleaning without special tools
During cleaning cycle, the electronically controlled purge valve provides a burst of ambient air that rapidly clears any accumulated moisture from the pumpheads out through the discharge port.
Spécifications
- Catalog No:
- 46620-636
- 46620-634
- 46620-632
- 46620-630
- Model:
- N842.3FT.40P
- N860.3FT.40P
- N840.3FT.40P
- N820.3FT.40P
- Vacuum:
- 3 torr
- 3 torr
- 8 torr
- 8 torr
- Protection class:
- IP 44 enclosure rating
- IP 44 enclosure rating
- IP 44 enclosure rating
- IP 44 enclosure rating
- W×D×H:
- 341×189×242 mm
- 331×291×278 mm
- 341×189×239 mm
- 312×177×220 mm