CrystalClear™ Crystallizing Dishes, without Spout
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Sturdy crystallizing dishes used for heating/cooling baths, crystallization and evaporation.
- Flat bottom and sides
- Autoclavable at 121 °C
- Strain Point: –515 °C
- Annealing Point: 565 °C
- Softening Point: 820 °C
Designed for repeated laboratory use, CrystalClear™ crystallizing dishes are made from industrial grade borosilicate 3.3 glass. Dishes are autoclavable, with excellent chemical resistance, a low temperature gradient, and can withstand high temperatures and thermal shocks. Reinforced rims are fire-polished to reduce chipping. Include printed marking spots.
Customers Who Bought This Also Bought