A propos de cet article
Ready to use GC TEMPase 2X Master Mixes I and II are designed for amplification of GC-rich sequences.
- High success rate with amplification of GC-rich DNA
- High sensitivity, specificity and product yield
- Diminished formation of non-specific products
- Reaction set-up at room temperature
- Processes up to 4 kb
The GC TEMPase 2X master mixes consist of TEMPase Hot Start DNA Polymerase combined with an optimized GC buffer system (GC buffer I or II), dNTPs and MgCl2. Just add DNA template and primers to carry out PCR.
GC-Rich DNA Target Kit consists of TEMPase Hot Start DNA polymerase and seperate tubes with GC buffer I and II. The kit is designed for initial testing before using one of the GC-TEMPase 2X Master Mixes.
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.