BCR® Cleanroom Face Masks, Berkshire
Fournisseur: Berkshire
Certificats
A propos de cet article
Berkshire masks are designed to protect your cleanroom or controlled environment from contamination.
- Filter Media: Meltblown polypropylene provides superior particle and bacterial filtration efficiency
- Outer layer: Blue polypropylene spunbond identifies the outside of the mask
- Inner layer: White polypropylene spunbond is smooth and non-irritating
- Nosepiece: Fully enclosed vinyl covered steel is adjustable to reduce 'blow by'
- Earloops: Knitted nylon spandex are soft and comfortable for long wear
- Ultrasonic bonding reduces contamination and outgassing
People are a significant source of contamination in any critical manufacturing process. Berkshire masks are designed to protect your cleanroom or controlled environment from human contamination.
Customers Who Bought This Also Bought