Axygen® 96-Well Assay Plates, 'V' Well, Corning (NZ onlyl)
Supplier: Corning
Certificates
About this item
Axygen® assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.
- SBS footprint
- Ultra-flat surface ensures proper sealing with heat sealing film
- Certified free of RNase, DNase and pyrogens
Specifications
- Catalog No:
- 47744-002
- 47744-004
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Working well volume:
- 500 µL for Round Bottom Plates, 500 µL for V-Bottom Plates
- 500 µL for Round Bottom Plates, 500 µL for V-Bottom Plates
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa