Corning® BioCoat® Poly-L-Ornithine/Laminin Plates, Corning
Fournisseur: Corning
Certificats
A propos de cet article
For some applications, the use of a combination of ECM proteins, such as Laminin (LM) and Fibronectin (HFN) or LM and attachment factors such as Poly-D-Lysine (PDL) or Poly-L-Orinthine (PLO) has been shown superior to the use of either alone. Corning BioCoat PDL/LM and PLO/LM Cultureware is suitable for culturing many different types of Peripheral Nervous System (PNS) and Central Nervous System (CNS) networks and is useful for promoting neural cell attachment and differentiation. Corning BioCoat LM/HFN Cultureware provides an in vitro environment that promotes cell attachment and extensive process formation.
- Enhancement of neuronal cell attachment to plastic and glass
- Promotion of neurite outgrowth
- Culture of glial cells as a feeder layer for neurons
- Construction of neural cell model systems to study CNS function, development, and diseases
- Storage: 2 to 8 °C, do not freeze
Customers Who Bought This Also Bought