Corning® Standard Polystyrene Microplates, 1536-well
Supplier: Corning
Certificates
About this item
These PS microplates conform to standard microplate footprint and dimensions, and are available in solid black or white, with round or flat bottoms, or in black clear bottom formats.
- Total well volume of 10 μl for round well plates and 12.8 μl for flat bottom plates
- Recommended working volume of up to 8 μl
- Round well bottoms for reduced air entrapment and improved CVs and Z factor
- Raised well bottoms for higher sensitivity
- Lids available separately
* NBS™ = Non binding surface
Specifications
- Catalog No:
- 71001-348
- 71001-346
- 71001-352
- 71001-350
- 89130-726
- 89130-730
- 89089-730
- 89089-732
- 89130-734
- 89089-746
- 89089-738
- 89089-742
- 89126-918
- 89126-916
- 66025-654
- 77776-918
- 66025-652
- 77776-916
- 89089-724
- 30621-094
- 89218-982
- 89089-728
- 89218-980
- 89218-978
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk