VWR® 2D Platform Rockers
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® 2D platform rockers are ideal for use in blotting and staining applications as well as gentle mixing of solutions and reagents.
- Space saving design and robust construction provide durability, reliability and quiet operation
- Large platforms with hooks for rubber strips easily accommodates many tubes, flasks and bottles
- Models feature adjustable speed and timer
- 2D large capacity rocker features adjustable tilt angle
- All models suitable for use in coldrooms and standard incubator environments
- 2 year replacement warranty
Certifications: CE Certified
Delivery: Units supplied with dimpled rubber mat (76595-818), and rubber strips 6/pack (76595-816), instruction manual and power cord.
Customers Who Bought This Also Bought