VWR® Tube Revolvers
Fournisseur: VWR International
Certificats
A propos de cet article
Efficient, powerful tube revolvers fixed or variable speed, with 5 interchangeable rotor series.
- Choose from fixed speed, 25 rpm or variable speed, 10 to 40 rpm models
- Variable speed model with LED display can be programmed to oscillate at every 40⁰ to simulate hand mixing with rotation and shaking
- Includes 5 interchangeable rotisseries to accommodate a variety of tube sizes simultaneously, each paddle can be adjusted from 0 to 90⁰
- Ergonomically designed for simple operation and minimal bench space usage
- Maintenance free motor is powerful and quiet
- 2-year replacement warranty
Four paddles or one 50 ml paddle can be attached to each unit simultaneously, increasing flexibility and efficiency.
Certifications: CE certified
Delivery: Units supplied with 2×10/15 ml paddles, 2×5/7 ml paddles, 2×1.5/2.0 ml paddles, 2×0.5/0.8 ml paddles, 1×50 ml paddle, instruction manual, power cord. Replacement paddles sold separately.
Customers Who Bought This Also Bought