VWR® Tube Rotators
Supplier: VWR International
Certificates
About this item
VWR® Tube rotators are ideal for applications requiring highly efficient and consistent mixing. VWR offers rugged and reliable fixed speed, digital and large capacity models.
- Adjustable tilt angle to accommodate different applications
- Variety of VWR® tube holders, carousels and drums available to accommodate various size sample tubes efficiently
- All units are suitable for use in cold room and incubator environments
- Digital tube rotator and the large capacity tube rotator offer adjustable speed, continuous and timer modes, with automatic stop
- 2 Drums available for 14 or 18 mm culture tubes for the large capacity tube rotator
- 2 year replacement warranty
Ideally suited for clinical, research, and academic applications.
Certifications: CE certified
Delivery: Units 76595-826 and 76595-826 supplied with VWR® carousel for 12×50 ml tubes (76595-856), large capacity model 76595-830 is suplied with carousel for 15x50 ml tubes (76595-862) plus instruction manual and power cord.
Customers Who Bought This Also Bought