PYREX® Disposable Culture Tubes, Rimless, Dispenser Pack and Bulk Pack, Corning
Supplier: Corning
Certificates
About this item
Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed.
Frequently Bought Together