PureAmp PCR and ELISA Plate Sealing Films
Fournisseur: MTC BIO, INC.
Certificats
A propos de cet article
A versatile sealing film for standard ELISA and PCR 96-well plates.
- RNase, DNase, DNA, and PCR inhibitor free
- EZ-Peel adhesive for spill-free removal
- Inhibits evaporation across all 96 wells (≤5%)
- Suitable for a wide temperature range
The sealing options in the PureAmp™ line include clear plastic film for end-point PCR, two optically clear films for qPCR, plus one pierceable aluminum membrane for storage and/or PCR. All PureAmp™ sealing products are pre-cut for a perfect fit on the plate every time and are guaranteed to keep evaporation rates under 5% in typical PCR applications. All PureAmp™ tubes, strips and plates are made of medical grade heat-resistant polypropylene. All PureAmp™ products are PCR certified and are guaranteed to be free of RNase, DNase, DNA, pyrogens, endotoxins and PCR inhibitors.
Packaging: Product is packaged in clear zip lock bags of 10 membranes within the standard pack of 100 membranes.
Customers Who Bought This Also Bought