DiluPhotometer™ OD600 for the Determination of Cell Density and Bradford Assays, Implen
# de catalogue CA10770-434
Fournisseur: IMPLEN, INC. CA
Spécifications
- Description:OD600 DiluPhotometer™ for the determination of cell density and Bradford assays
- Cat. no.:CA10770-434
- Supplier No.:OD600-10
Spécifications
A propos de cet article
Fast and easy determination of cell density and 600 nm.
- Small, portable spectrophotometer
- Quantify protein with Bradford protein assay
- Operating wavelength is 590 to 610 nm
Small, easy to use instrument for the determination of cell density and Bradford assays in wavelengths ranging from 590 to 610 nm. Battery operated.
Delivery: Charger included; includes 1 pack of DiluCell 10 cuvettes.
Customers Who Bought This Also Bought