TDE Series Ultra-Low Freezers with Chart Recorder, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
TDE sereies ultra-low temperature freezers are part of the Standard Performance (STP) family.
Insulation: water-blown polyurethane foam
Certifications: cULus; FDA Class II.
Packaging: Pack includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed chart recorder.
Spécifications
- Catalog No:
- 76595-626
- 76595-642
- 76595-586
- 76595-616
- 76595-568
- 76595-634
- 76595-594
- 76595-578
- Model:
- TDE60086FDRCR
- TDE60086LDRCR
- TDE50086LARCR
- TDE60086FARCR
- TDE50086FARCR
- TDE60086LARCR
- TDE50086LDRCR
- TDE50086FDRCR
- Alarm:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Compressor:
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- Electrical:
- 208 - 230 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 115 V, 60 Hz
- 115 V, 60 Hz
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 208 - 230 V, 60 Hz
- Door type:
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- Material:
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Plug:
- NEMA 6-15
- NEMA 6-15
- NEMA 5-20
- NEMA 5-20
- NEMA 5-20
- NEMA 5-20
- NEMA 6-15
- NEMA 6-15