TDE Series Ultra-Low Freezers with Racks and Boxes, −40 °C, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
TDE series −40 °C ultra-low temperature freezers pack includes one ULT freezer and two shelves of sliding drawer racks and boxes.
Certifications: EPA SNAP, Class II.
Spécifications
- Catalog No:
- 76595-556
- 76595-520
- 76595-602
- 76595-504
- 76595-512
- 76595-496
- Model:
- TDE50040FARK
- TDE40040LDRK
- TDE60040FARK
- TDE40040FDRK
- TDE40040LARK
- TDE40040FARK
- Alarm:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Compressor:
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- Electrical:
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 115 V, 60 Hz
- Door type:
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- Material:
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Plug:
- NEMA 5-20
- NEMA 6-15
- NEMA 5-20
- NEMA 6-15
- NEMA 5-20
- NEMA 5-20