TDE Series Ultra-Low Freezers with Racks and Boxes, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
TDE series ultra-low temperature freezers are part of the Standard Performance (STP) family.
Certifications: cULus; FDA Class II.
Packaging: Pack includes one ULT freezer and two shelves of sliding drawer racks and boxes.
Spécifications
- Catalog No:
- 76595-644
- 76595-490
- 76595-488
- 76595-486
- 76595-628
- 76595-482
- 76595-588
- 76595-548
- 76595-580
- 76595-570
- 76595-618
- 76595-596
- 76595-636
- 76595-542
- 76595-536
- 76595-528
- Model:
- TDE60086LDRK
- TDE30086LDRK
- TDE30086LARK
- TDE30086FDRK
- TDE60086FDRK
- TDE30086FARK
- TDE50086LARK
- TDE40086LDRK
- TDE50086FDRK
- TDE50086FARK
- TDE60086FARK
- TDE50086LDRK
- TDE60086LARK
- TDE40086LARK
- TDE40086FDRK
- TDE40086FARK
- Alarm:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Compressor:
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- Electrical:
- 208 - 230 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- Door type:
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- Material:
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Plug:
- NEMA 6-15
- NEMA 6-15
- NEMA 5-20
- NEMA 6-15
- NEMA 6-15
- NEMA 5-20
- NEMA 5-20
- NEMA 6-15
- NEMA 6-15
- NEMA 5-20
- NEMA 5-20
- NEMA 6-15
- NEMA 5-20
- NEMA 5-20
- NEMA 6-15
- NEMA 5-20