TDE Series Ultra-Low Temperature Freezers with CO₂ Back-up System, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
TDE series ultra-low temperature freezers is part of the Standard Performance (STP) family.
Insulation: water-blown polyurethane foam
Certifications: cULus; FDA Class II.
Packaging: pack includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed CO₂ back-up system.
Spécifications
- Catalog No:
- 76595-526
- 76595-534
- 76595-624
- 76595-546
- 76595-584
- 76595-640
- 76595-566
- 76595-614
- 76595-576
- 76595-592
- 76595-540
- 76595-632
- Model:
- TDE40086FARCO
- TDE40086FDRCO
- TDE60086FDRCO
- TDE40086LDRCO
- TDE50086LARCO
- TDE60086LDRCO
- TDE50086FARCO
- TDE60086FARCO
- TDE50086FDRCO
- TDE50086LDRCO
- TDE40086LARCO
- TDE60086LARCO
- Alarm:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Compressor:
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- H-Drive compressor
- Electrical:
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 208 - 230 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 115 V, 60 Hz
- 208 - 230 V, 60 Hz
- 208 - 230 V, 60 Hz
- 115 V, 60 Hz
- 115 V, 60 Hz
- Door type:
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- 1 outer, 4 inner
- Material:
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Plug:
- NEMA 5-20
- NEMA 6-15
- NEMA 6-15
- NEMA 6-15
- NEMA 5-20
- NEMA 6-15
- NEMA 5-20
- NEMA 5-20
- NEMA 6-15
- NEMA 6-15
- NEMA 5-20
- NEMA 5-20