A propos de cet article
The Mopec prostate biopsy kits come ready to use in a set of 8, 12, or 16 containers. Containers are 20 ml capacity, half-filled with 10% neutral buffered formalin, and feature a double-lock closure that guarantees the container’s seal is fully secured.
- 20 ml containers are half-filled with 10% neutral buffered formalin
- Feature a double-lock closure that guarantees the container’s seal is fully secured
- Leak-resistant seal on containers are 95 kPa compliant
- Contains specific instructions for use and a cardboard insert labeled for placement of the appropriate prostate section
- Sold individually
Each kit contains specific instructions for use and a cardboard insert labeled for placement of the appropriate prostate section.