Formaway Waste Collection Container, Mopec
Fournisseur: Mortech Manufacturing
Certificats
A propos de cet article
Mopec’s Formaway collection containers are used to collect spent formalin for neutralizing.
- Collect waste in a designated collection container
- Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste
- Add the entire contents of one formaway bottle to each gallon of aldehyde waste
- Secure lid on the collection container and agitate container to thoroughly mix solutions, make sure the agitation ultimately results in complete dissolution of formaway formalin neutralizer powder
- Allow the mixed solution to stand for 20 to 25 minutes for the reaction to be complete
- If required, test treated solution for any aldehyde residual prior to disposal (follow instructions for formaway formaldehyde residual screening test kit)
- Pour treated waste into sanitary sewer and rinse container with cold tap water, preparing it for re-use
Sold individually.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.