Eisco Drechsel Gas Washing Bottles
Fournisseur: Wards
Certificats
A propos de cet article
Remove impurities from gases.
- Made from high quality borosilicate 3.3 glass
- Interchangeable ground glass joint allows for easy sealing and exchange
- Both bottle and joint head are autoclavable
Autoclavable, with superior chemical durability. Low temperature gradient; can withstand high temperatures and thermal shocks.
Customers Who Bought This Also Bought