Eisco Polypropylene Aspirator Bottle with Leak Proof Spigot
Supplier: Wards
Certificates
About this item
Two study handles molded right into the bottle for ease of carrying or moving.
- Durable bottles for storing and dispensing laboratory solutions
- Rigid polypropylene construction provides excellent chemical resistance to acids, alcohols and alkalis, and very good resistance to aldehydes and hydrocarbons
- Equipped with threaded cap with gasket and leak proof threaded spigot with on/off indicator, and stopcock
When autoclaving, threads should be totally disengaged on spigot.
Customers Who Bought This Also Bought