Axygen® Automation Tips for PerkinElmer, Zymark® RapidPlate®, Sciclone®
Fournisseur: Corning
Certificats
A propos de cet article
Transparent PP.
- Sterile tips: Sterilised by gamma radiation
- Filter tips: PE filter prevents any pipette contamination
- Free of RNase, DNase, DNA and pyrogens
Available options:
- With or without aerosol-resistant filters
- Nonsterile or sterile
- Standard or Axygen® Maxymum Recovery® surface
Customers Who Bought This Also Bought