Evergreen 2 Compartment Cassette, Caplugs Evergreen
# de catalogue 76502-840
Fournisseur: Caplugs
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Color:White
- Description:Acetal cassettes
- Cat. no.:76502-840
- No. of Compartments:2
Spécifications
A propos de cet article
Caplugs Evergreen two compartment cassettes with 0.6 mm holes can help you customize your biopsy processing with added efficiency and economy.
- High density acetal construction
- Acetal is resistant to histological solvents such as alcohol and xylene
- Acetal will also keep the specimen submerged
- Modified ball-and-socket hinge design for ease of opening and closing and also for detachment
- Lid and base preassembled
Customers Who Bought This Also Bought