Skolnik UN Certified Steel Open Head Drums with Bolt Ring Closure
Fournisseur: Qorpak
Certificats
A propos de cet article
Qorpak® Skolnik UN certified steel drums are thicker, heavier, and stronger than industry standards require, offer extreme durability, and comply with domestic and international regulations.
- Meets the rigorous UN packaging requirements of 49 CFR
- Tested per the DOT and UN packaging test requirements
- Heavier and stronger than industry standards
- Better endures nail and forklift punctures than thinner drums
Do not risk place your volatile, expensive, irreplaceable, hazardous, or precious contents in any other vessel. Protect your products, reputation, and the environment with a superior quality drum.
Certifications: UN certified for solids and/or liquids.
Customers Who Bought This Also Bought