COSA™ CIP 96 Alkaline Cleaner
Fournisseur: ECOLAB LIFE SCIENCES
Certificats
A propos de cet article
COSA™ CIP 96 is a strong alkaline, surfactant-free cleaner.
- Surfactant-free formula for ease of rinsing.
- Chemical concentration can be dosed and monitored using conductivity.
- With the addition of cleaning boosters in the COSA™ line, COSA™ CIP 96 is suitable for cleaning product lines with fatty and gel containing residues.
- Supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation.
COSA™ CIP 96 is a strong alkaline, surfactant-free cleaner for cleaning in the pharmaceutical and personal care industries.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.