Quantum Levitation Force Experiment
# de catalogue 470346-494
Fournisseur: QUANTUM LEVITATION LTD
Spécifications
- Description:Quantum level forces experiment
- Cat. no.:470346-494
Spécifications
A propos de cet article
Explore the magnetic forces between magnets, superconductors and metals.
- The Meissner effect
- Flux pinning
- Magnetic forces
- Letz' law
Visualize and measure the magnetic forces that Quantum Levitation so unique and unintuitive - see how a superconductor is repelled by a magnet (positive force - Meissner effect) and after it has been locked, it is attracted (negative forces - flux pinning) to it.
Customers Who Bought This Also Bought