Universal U-Electrode, Mettler-Toledo
# de catalogue 76580-208
Fournisseur: Mettler Toledo
Spécifications
- Description:Universal U-Electrode
- Cat. no.:76580-208
Spécifications
A propos de cet article
Anti-static solution for samples and tare containers with dimensions of 12×25 cm.
- The U-electrode can be placed next to the balance, and will remove the static charge to avoid inaccurate weighing results
Customers Who Bought This Also Bought