Spectrophotometers, Easy UV/Easy VIS, Mettler-Toledo
Supplier: Mettler Toledo
Certificates
About this item
The Easy UV and Easy VIS spectrophotometers offer 3-in-1 functionality and is designed to greatly facilitate routine spectroscopic workflows.
- Save space – The compact design similar to an A4 paper reduces clutter on your lab bench
- Enjoy fast, compliant color measurement – Includes 30 color scales (e.g. CIELAB, APHA, Gardner, and more.)
- Avoid errors – PathDetect™ securely checks the path length so your results are accurate
- Test water quickly – The SQKitReader™ accessory unlocks over 150 Spectroquant® water test kit methods
- Access methods easily – Built-in features such as a favorite list and LongClick™ speed up routine analyses
- Clean up quickly thanks to a washable drip tray
- The EasyPlus spectrophotometers are designed to reduce the effects of turbidity on measurements (please contact your local representative for culturing applications)
The instrument is simple and easy-to-use, and ensures accurate, repeatable measurements. Built-in methods also eliminate calculation and transcription errors.
Certifications: CE, CSA.
Specifications
- Catalog No:
- 76580-224
- 76580-222
- Display:
- 4.3" touchscreen
- 4.3" touchscreen
- Photometric accuracy:
- ±0.005A
- ±0.005A
- Detector:
- Dual silicon photodiodes
- Dual silicon photodiodes