VWR® Cleanroom Latex Gloves, Class 10, Non Sterile
Fournisseur: VWR International
Certificats
A propos de cet article
Ambidextrous, powder-free gloves designed to provide comfort and tactile sensitivity.
- Ambidextrous, beaded cuff
- Powder-free, silicone-free
- Natural rubber latex
- Low particle count
- Specialized manufacturing process
Latex, nonsterile cleanroom gloves are available in Class 10 designation.
Spécifications
- Catalog No:
- 76579-676
- 76579-674
- 76579-672
- 76579-678
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile