ContactUsLinkComponent
CAE® Lucina Maternal and Fetal Trainer
CAE® Lucina Maternal and Fetal Trainer
# de catalogue CA470346-840
SIMULATOR LUCINA- LIGHT
CAE® Lucina Maternal and Fetal Trainer
# de catalogue CA470346-840
Spécifications
  • Description:
    Lucina
  • Cat. no.:
    CA470346-840
Spécifications
A propos de cet article
A better childbirth simulator that's beyond exceptional.
  • Two adult female patients. One patient simulator
  • Beauty meets functionality
  • Advanced programming for life-saving skills training
  • Impressive library of scenarios
CAE lucina is the only wireless childbirth simulator with validated, integrated maternal-fetal physiology and interchangeable static cervices to train on all the stages of delivery and the rare emergency scenario. Lucina brings an impressive blend of features to the market including a more realistic and controllable birthing process, better articulation for labor and delivery maneuvers, and predicted APGAR scores based on the integrated maternal-fetal physiology. From normal delivery and breech birth to shoulder dystocia and eclampsia, lucina does it all. With 10 pre-configured Simulated Clinical Experiences (SCEs), lucina allows instructors to increase throughput and manage multiple demands effortlessly.
Lucina is a hybrid solution for both obstetrical patient management training and non-gravid, female patient care education. Tuck the simulated fetus into the delivery abdomen and lucina is ready for ob rounds. Realistic palpable uterine contractions, a reliable birthing mechanism supporting automatic descent, and crowning and rotation of the fetus provide true-to-life learning experiences. Advanced cpr metrics measure the consistency of compressions and ventilations, as well as coronary and cerebral perfusion. Hand placement, chest recoil, and left lateral tilt are detected and logged by the müse operating system. 10 pre-configured simulated clinical experiences (sce's) that come ready to operate, from normal vaginal delivery to urgent and interventional birthing maneuvers (including leopold's maneuver, zavanelli maneuver, trendelenburg position, shoulder dystocia, vertex and breech delivery).
Customers Who Bought This Also Bought