Hydraulic Ram Discharge System, MorehouseCowles
# de catalogue 76553-868
Fournisseur: MorehouseCowles
Spécifications
- Description:Hydraulic Ram discharge system, 1 gal. tank
- Cat. no.:76553-868
Spécifications
A propos de cet article
Your heavy, viscous products can be effortlessly removed from the tank when you activate the MorehouseCowles hydraulic Ram discharge system. With a powerful push, the product is forced out of the dispersion tank, so it takes less labor and time to empty and clean the equipment.
- Avoids product waste
Delivery: Comes completely assembled and ready to use.
Packaging: Shipped in a wooden crate.
Customers Who Bought This Also Bought