VWR® Plus Speciality Laboratory Refrigerators with Glass Door
Fournisseur: Horizon Scientific
Certificats
À propos de cet article
These VWR® Plus Series laboratory refrigerators offer premium cold storage with superior temperature control.
- Microprocessor temperature controller
- Digital temperature display and alarm module
- Audible, visual, senor error, and power failure alarms
- Alarm validation and alarm mute with ring back
- Keyed door lock
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These speciality glass door laboratory refrigerators offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, probe access, and precise temperature monitoring through microprocessor control. The Plus Series line includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Material: White powder coat
Certifications: UL/C-UL listed.
*Select models ENERGY STAR certified: 76580-152, 76580-156
*Select models ENERGY STAR certified: 76580-152, 76580-156
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Spécifications
- Catalog No:
- 76580-160
- 76580-164
- 76580-152
- 76580-156
- Environmentally Preferable:
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Cat. No.:
- 76580-160
- 76580-164
- 76580-152
- 76580-156
- Defrost:
- Cycle
- Cycle
- Cycle
- Cycle
- Shelves/drawers:
- 8 adjustable shelves (adjustable in 1/2" increments)
- 12 adjustable shelves (adjustable in 1/2" increments)
- 4 adjustable shelves (adjustable in 1/2" increments)
- 4 adjustable shelves (adjustable in 1/2" increments)
- Door type:
- 2 glass doors
- 3 glass doors
- 1 glass door, right hinged
- 1 glass door, right hinged
- Electrical:
- 115 V, 60 Hz, 8.5 A, ½ HP, BTU/HR 7315
- 115 V, 60 Hz, 8.5 A, ½ HP, BTU/HR 7315
- 115 V, 60 Hz, 5.4 A, ⅓ HP, BTU/HR 4932
- 115 V, 60 Hz, 5.4 A, ⅓ HP, BTU/HR 4932
- Weight:
- 225 kg (495 lbs.)
- 313 kg (690 lbs.)
- 134 kg (295 lbs.)
- 156 kg (345 lbs.)
- Environmental certification:
- Uses Natural Hydrocarbon refrigerants
- Uses Natural Hydrocarbon refrigerants
- Uses Natural Hydrocarbon refrigerants
- Uses Natural Hydrocarbon refrigerants