SureSTART™ High Recovery Glass Screw Top 1.7 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Choose Thermo Scientific™ SureSTART™ 1.7 ml high recovery glass screw top microvials, performance Level 3, when you need to maximize the injection volume for <2 ml samples.
With outer dimensions of 12×32 mm, these clear or amber glass microvials have an internal conical reservoir that provides a total usable volume of 1.5 ml and a residual volume of 4 µl. This is a Performance Level 3 vials, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyzes.
Customers Who Bought This Also Bought