PUREGRIP® Aspirator Bottles, with Top Thread and GL 45 Caps, Foxx Life Sciences
Fournisseur: Foxx Life Sciences
Certificats
A propos de cet article
PUREGRIP® Aspirator Bottles feature a top thread and GL 45 caps, and they have a tubular sidearm outlet near the bottom for attachment of flexible tubing. Bottles are very useful as a delivery/storage container for solutions.
- Sizes range from 250 ml to 20 L
- Highest Polypropylene cap temperature rating in the industry, 142 °C
- Industry standard GL45 thread, compatible with Autofil™ Bottle-Top Filters and EZBio®Single-Use Cap Assemblies
- Large, visible writing surface on bottle for additional labeling
- White graduations for identification of approximate volume
- Retrace code for identification
- BOROSIL® 3.3 Borosilicate glass is mechanically strong, chemically resistant, and withstands thermal shock
- Bottle, VersaCap®, and pouring ring are autoclavable, and comply with USP-VI
These bottles feature sturdy ring necks with heavy-duty rims. They can withstand repeated sterilization (wet or dry).
Certifications: Complies with IS 1388 (Part III), EN ISO 4796-3.
Customers Who Bought This Also Bought