EZBio® Media Bottles, Square Storage Bottles, with and without Caps, Foxx Life Sciences
Fournisseur: Foxx Life Sciences
Certificats
A propos de cet article
Foxx Life Sciences offers the widest range of EZBio® Single Use Media Bottles on the market, with a variety of sizes, cap options, and materials available.
- Media bottles are available in five different sizes: 60 ml, 125 ml, 250 ml, 500ml, 1 L, 2 L
- Made entirely of 100% USP Class VI components from the United States
- Gamma Irradiated in the United States
- EZBio® Single Use Media Bottles are compatible with the EZBio® cap assemblies
With a leakproof design and revolutionary VersaCap® technology, Foxx offers the most cost effective product on the market. EZBio® Bottles can be used for sampling, sorting media, biologicals, and many other aqueous solutions.
Certifications: USP Class VI material.
Customers Who Bought This Also Bought