Support Rings with Opening, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Easily move separatory funnels wit this open ring design.
- Open support rings
- Ideal for use with funnels, boiling flasks, and separatory funnels
Steel rod, zinc-plated extension support rings. Rod diameter 0.27" (7 mm) and the straight rod portion of each is 7.25" (184 mm) long.
Frequently Bought Together