Chalkboard Drawing Instrument, Triangle, 60°, United Scientific Supplies
# de catalogue 470340-120
Fournisseur: United Scientific Supplies
Spécifications
- Description:Chalkboard drawing instrument, triangle, 60°
- Cat. no.:470340-120
Spécifications
A propos de cet article
Oversized triangle for whiteboards.
- 60° triangle with 22" base
- Made of white polystyrene
- Features a handle for ease of use
Durable, lightweight oversized plastic triangle, designed for whiteboards and chalkboards.
Customers Who Bought This Also Bought