Chalkboard Drawing Instrument, Triangle, 45°, United Scientific Supplies
# de catalogue 470339-352
Fournisseur: United Scientific Supplies
Spécifications
- Description:Chalkboard drawing instrument, triangle, 45°
- Cat. no.:470339-352
Spécifications
A propos de cet article
Oversized triangle for whiteboards.
- 45° triangle with 23" base and 16" sides
- Made of white polystyrene
- Features a handle for ease of use
Durable, lightweight oversized plastic triangle, designed for whiteboards and chalkboards.
Customers Who Bought This Also Bought