Chalkboard Drawing Instrument, Meter Ruler, United Scientific Supplies
# de catalogue 470339-380
Fournisseur: United Scientific Supplies
Spécifications
- Description:Chalkboard drawing instrument, meter ruler
- Cat. no.:470339-380
Spécifications
A propos de cet article
Oversized ruler for whiteboards.
- Meter ruler, graduated in inches (1 to 39")
- Made of white polystyrene
- Features a handle for ease of use
Durable, lightweight oversized plastic ruler, designed for whiteboards and chalkboards.
Customers Who Bought This Also Bought