Compact Glove Boxes, Yamato
Fournisseur: Yamato Scientific
Certificats
A propos de cet article
Compact, completely sealed glove box with transfer chamber and flat access door for basic isolation and/or containment.
- High visibility interior, cleanliness, safety, complete containment, portability, and a draft-free atmopshere
- Completed systems, nothing needs to be added except gas of choice: nitrogen, argon, or other inert gases
- Installed purging valves are ideal for lowering oxygen and humidity levels
Very useful when working with toxic substances, asbestos, fibers, sewage residue, and harmful liquid vapors. Made in the USA.
Certifications: CE certification.
Customers Who Bought This Also Bought