Eisco Copper Hemispherical Water Bath
Fournisseur: Wards
Certificats
A propos de cet article
Provides maximum volume at shallow depth.
- Lid is comprised of a set of concentric rings
- Features two side handles and a top handle for safe and easy handling
Hemispherical water bath made of copper sheet, which is ideal for the best heat conductivity.
Customers Who Bought This Also Bought