Eisco Porcelain Crucible with Lid, Short Form
Fournisseur: Wards
Certificats
A propos de cet article
High thermal performance and enhanced chemical resistance.
- Crucible for basic laboratory use to heat chemical compounds
- Short form body with lid
- Constructed of high quality glazed porcelain for strength and durability
- Can withstand temperatures up to 1050 °C (1900 °F)
Recommended rate of heating and cooling should NOT exceed 200 °C per hour.
Customers Who Bought This Also Bought