Eisco Glass Aspirator Bottle with Ribbed Outlet
Fournisseur: Wards
Certificats
A propos de cet article
Ungraduated glass aspirator bottles have a ribbed sidearm outlet located at the base of the bottle for attaching tubing and an open, beaded neck.
- Made of high-quality, heavy-duty, borosilicate 3.3 glass
- Superior durability and chemical resistance
- Can withstand high temperatures and thermal shocks
- Integral ribbed outlet at base of bottle for attaching tubing
Aspirator bottles are autoclavable, and offers superior chemical durability and a low temperature gradient. They can withstand high temperatures and thermal shocks.
Eisco borosilicate glass has the following heat tolerances: strain point of 515 °C, annealing point of 565 °C, and softening point of 820 °C.
Customers Who Bought This Also Bought