United Scientific™ Spot Plate, Porcelain, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Glazed porcelain spot plate.
- Color-reaction or spot plate
- Crafted from porcelain; glazed except for bottom surface
- Autoclavable
Color-reaction or spot plate with three wells. Crafted from porcelain; glazed except for bottom surface.
Customers Who Bought This Also Bought