96 Deep-Well Plates, Sterile, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Medical grade deep well and comb.
- Made of medical-grade polypropylene (PP)
- Uniform, flat upper plate uniform, easy to seal, uniform aperture
- Stackable, easy to store
The product is made of medical grade PP with flatness of the upper surface being controllable within 0.25 mm and is easy for film sealing.
Customers Who Bought This Also Bought