United Scientific™ DNA Model Kits With Four Models And CD, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Popular model for investigating DNA.
- Classroom kit includes one preassembled DNA model, built to a scale of 10cm to 1nm, and four unassembled models
- Central pillar support allows assembled model to rotate on circular stand
- Each model can be used by a group of up to 5 students
- Instructional CD explains key concepts related to DNA structure and includes step-by step instructions for model assembly
- Contains small parts; not for children under 3 years
DNA Model Kit includes one preassembled DNA model, four unassembled models, and one instructional CD. The CD explains key concepts related to DNA structure and includes step-by step instructions for assembly of the DNA Models.
Model is built to a scale of 10 cm to 1 nm and accurately depicts a 16 base pair section of DNA. The model is supported by a central pillar which can rotate on a circular stand.
Each model can be used by a group of 5 students. Kit contains no consumable parts and is designed for repeated use.
Caution: Contains small parts; not for children under 3 years.
Customers Who Bought This Also Bought