Chalkboard Drawing Instrument, Protractor, 180° , United Scientific Supplies
# de catalogue 470340-110
Fournisseur: United Scientific Supplies
Spécifications
- Description:Chalkboard drawing instrument, protractor, 180°
- Cat. no.:470340-110
Spécifications
A propos de cet article
Oversized protractor for whiteboards.
- 180°, marked at 1° intervals
- Made of white polystyrene
- Features a handle for ease of use
Durable, lightweight oversized plastic protractor, designed for whiteboards and chalkboards.
Customers Who Bought This Also Bought