Cryo Cube Boxes, PP, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Polypropylene cryo boxes with alpha-numeric grid.
- Autoclavable
- Temperatures range: –90 °C to +121 °C.
Manufactured of polypropylene, these autoclavable boxes are stackable for sample storage in 1.5 to 2.0 ml microtubes and 1.0 to 2.0 ml cryogenic vials. Boxes feature a hinged lid connected to the base and a locking system to ensure secure closure. A molded alphanumeric grid allows for easy identification of samples. Dimensions of cryo boxes are designed to be compatible with vertical, horizontal, and drawers style freezer racks. Each pack includes two yellow boxes and two blue boxes.
Customers Who Bought This Also Bought