Porcelain Casserole, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Porcelain dish with a handle.
- Autoclavable
- Features lip and flat handle
- Glazed inside and out, except for rim and top of handle
- Maximum use temperature of 1150 °C (2102 °F)
Porcelain casserole featuring lip and flat handle. Excellent resistance to acids and alkalies, not including hydrofluoric acid. Autoclavable and glazed inside and out, except for rim and top of handle. Designed to withstand temperatures up to 1150 °C; to prevent thermal stress cracks, temperature change rate should no exceed 200 °C per hour.
Customers Who Bought This Also Bought